accessibility menu, dialog, popup

UserName

CAS RN 590-93-2

IUPAC Name: but-2-ynoic acid
Synonyms: 2-butynoic acid
Molecular Weight (g/mol): 84.07
Molecular Formula: C4H4O2
InChi Key: LUEHNHVFDCZTGL-UHFFFAOYSA-N
SMILES: CC#CC(O)=O
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)