missing translation for 'onlineSavingsMsg'
Learn More

Tetrolic Acid 98.0+%, TCI America™

Catalog No. T11991G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11991G 1 g
T11995G 5 g
T119925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T11991G Supplier TCI America Supplier No. T11991G

Chemical Identifiers

CAS 590-93-2
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.07
MDL Number MFCD00004363
InChI Key LUEHNHVFDCZTGL-UHFFFAOYSA-N
Synonym 2-butynoic acid, tetrolic acid, 3-methylpropiolic acid, 1-propynecarboxylic acid, 2-butynoicacid, tetrolsaure, butynoic acid, 2-butynic acid, propinecarboxylic acid
PubChem CID 68535
IUPAC Name but-2-ynoic acid
SMILES CC#CC(O)=O

Specifications

Melting Point 77°C
Color White-Yellow
Boiling Point 203°C
Quantity 1 g
UN Number 3261
Formula Weight 84.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrolic Acid