missing translation for 'onlineSavingsMsg'
Learn More

Tetratetracontane, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015317
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015317 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015317 Supplier Merck Emd Millipore Supplier No. 88144250MG
Only null left

Specifications

CAS 7098-22-8
Physical Form Neat
Linear Formula CH3(CH2)42 CH3
MDL Number MFCD00015268
Melting Point 85°C to 87°C; 85°C to 88°C
Type Analytical Standard
Quantity 250 mg
Formula Weight 619.19
Percent Purity ≥98% (GC)
Grade Analytical Standard