missing translation for 'onlineSavingsMsg'
Learn More

Tetrasodium Ethylenediaminetetraacetate Tetrahydrate 98.0+%, TCI America™

Catalog No. E0565500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0565500G 500 g
E056525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0565500G Supplier TCI America Supplier No. E0565500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13235-36-4 | C10H20N2Na4O12 | 452.23 g/mol

Chemical Identifiers

CAS 13235-36-4
Molecular Formula C10H20N2Na4O12
Molecular Weight (g/mol) 452.23
MDL Number MFCD00150027
InChI Key XFLNVMPCPRLYBE-UHFFFAOYSA-J
Synonym tetrasodium ethylenediaminetetraacetate tetrahydrate, edetate sodium tetrahydrate, unii-l13nhd21x6, ethylenediaminetetraacetic acid tetrasodium salt tetrahydrate, acmc-209bp2, ksc910c4l, edta, tetrasodium tetrahydrate salt, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, tetrasodium salt, tetrahydrate, ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, sodium salt, hydrate 1:4:4
PubChem CID 23287279
IUPAC Name tetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate tetrahydrate
SMILES O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Color White
Quantity 500 g
Formula Weight 380.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrasodium Ethylenediaminetetraacetate Tetrahydrate