accessibility menu, dialog, popup

UserName

CAS RN 13235-36-4

IUPAC Name: tetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate tetrahydrate
Synonyms: tetrasodium ion(4-) edta tetrahydrate
Molecular Weight (g/mol): 452.23
Molecular Formula: C10H20N2Na4O12
InChi Key: XFLNVMPCPRLYBE-UHFFFAOYSA-J
SMILES: O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)