missing translation for 'onlineSavingsMsg'
Learn More

Tetrasodium Ethylenediaminetetraacetate Dihydrate 98.0+%, TCI America™

Catalog No. p-7128837
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E009925G 25 g
E0099500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E009925G Supplier TCI America Supplier No. E009925G
Only null left

Chemical Identifiers

CAS 10378-23-1
Molecular Formula C10H20N2Na4O10
Molecular Weight (g/mol) 420.233
MDL Number MFCD00012460
InChI Key DGWOWDJDIBPYOR-UHFFFAOYSA-N
Synonym ethylenediaminetetraacetic acid tetrasodium salt dihydrate, tetrasodium ethylenediaminetetraacetate dihydrate, edta-na4, edta tetrasodium salt, edta tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate, 64-02-8 anhydrous, ksc909a7h, edta tetrasodium salt dihydrate, tetrapotassium ion 4-edta dihydrate
PubChem CID 131843362
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na]

Specifications

Melting Point 300°C
Color White
Quantity 25 g
Formula Weight 380.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrasodium Ethylenediaminetetraacetate Dihydrate
EINECSNumber (2)-1265
RTECSNumber AH5075000
TSCA Yes