accessibility menu, dialog, popup

UserName

NOHOOHONOHOOOHH2OH2ONaNaNaNa

CAS RN 10378-23-1

IUPAC Name: 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid dihydrate tetrasodium
Synonyms: edetic acid dihydrate tetrasodium
Molecular Weight (g/mol): 420.23
Molecular Formula: C10H20N2Na4O10
InChi Key: DGWOWDJDIBPYOR-UHFFFAOYSA-N
SMILES: O.O.[Na].[Na].[Na].[Na].OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (5)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Grade 
  • (1)
  • (5)
  • (2)
Physical Form 
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (5)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (5)
  • (4)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (2)
  • (5)
  • (3)