missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrapropylammonium Perruthenate, TCI America™
$334.98 - $974.13
Chemical Identifiers
| CAS | 114615-82-6 |
|---|---|
| Molecular Formula | C12H35NO4Ru |
| Molecular Weight (g/mol) | 358.485 |
| MDL Number | MFCD00074914 |
| InChI Key | HUCLFLGLPCVDMZ-UHFFFAOYSA-M |
| Synonym | 1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1- |
| PubChem CID | 127020979 |
| IUPAC Name | ruthenium;tetrapropylazanium;hydroxide;trihydrate |
| SMILES | CCC[N+](CCC)(CCC)CCC.O.O.O.[OH-].[Ru] |
Chemical Identifiers
| 114615-82-6 | |
| 358.485 | |
| HUCLFLGLPCVDMZ-UHFFFAOYSA-M | |
| 127020979 | |
| CCC[N+](CCC)(CCC)CCC.O.O.O.[OH-].[Ru] |
| C12H35NO4Ru | |
| MFCD00074914 | |
| 1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1- | |
| ruthenium;tetrapropylazanium;hydroxide;trihydrate |
Specifications
| 114615-82-6 | |
| Black-Green | |
| MFCD00074914 | |
| 1479 | |
| HUCLFLGLPCVDMZ-UHFFFAOYSA-M | |
| ruthenium;tetrapropylazanium;hydroxide;trihydrate | |
| 127020979 | |
| Crystalline Powder |
| 163°C | |
| C12H35NO4Ru | |
| 1 g | |
| 1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1- | |
| CCC[N+](CCC)(CCC)CCC.O.O.O.[OH-].[Ru] | |
| 358.485 | |
| 351.43 | |
| Tetrapropylammonium Perruthenate |
Safety and Handling
TSCA : No