accessibility menu, dialog, popup

UserName

CAS RN 114615-82-6

IUPAC Name: tetrapropylazanium tetrahydrate ruthenium
Synonyms: tetrapropylammonium perruthenate
Molecular Weight (g/mol): 359.49
Molecular Formula: C12H36NO4Ru
InChi Key: HUCLFLGLPCVDMZ-UHFFFAOYSA-N
SMILES: O.O.O.O.[Ru].CCC[N+](CCC)(CCC)CCC
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results