missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium Perruthenate, TCI America™

Catalog No. T15595G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15595G 5 g
T15591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15595G Supplier TCI America Supplier No. T15595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 114615-82-6
Molecular Formula C12H35NO4Ru
Molecular Weight (g/mol) 358.485
MDL Number MFCD00074914
InChI Key HUCLFLGLPCVDMZ-UHFFFAOYSA-M
Synonym 1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1-
PubChem CID 127020979
IUPAC Name ruthenium;tetrapropylazanium;hydroxide;trihydrate
SMILES CCC[N+](CCC)(CCC)CCC.O.O.O.[OH-].[Ru]

Specifications

Melting Point 163°C
Color Black-Green
Quantity 5 g
UN Number 1479
Formula Weight 351.43
Physical Form Crystalline Powder
Chemical Name or Material Tetrapropylammonium Perruthenate