missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium Hydroxide (20-25% in Water), TCI America™

Catalog No. T102025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T102025ML 25 mL
T1020500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T102025ML Supplier TCI America Supplier No. T102025ML

Chemical Identifiers

CAS 4499-86-9
Molecular Formula C12H29NO
Molecular Weight (g/mol) 203.37
MDL Number MFCD00009360
InChI Key LPSKDVINWQNWFE-UHFFFAOYSA-M
Synonym tetrapropylammonium hydroxide, tetra-n-propylammonium hydroxide, 1-propanaminium, n,n,n-tripropyl-, hydroxide, tpaoh, tetrapropylammonium oxide, unii-t686luy7nk, tetrapropylazanium hydroxide, t686luy7nk, ammonium, tetrapropyl-, hydroxide, n,n,n-tripropyl-1-propanaminium hydroxide
PubChem CID 20586
IUPAC Name tetrapropylazanium hydroxide
SMILES [OH-].CCC[N+](CCC)(CCC)CCC

Specifications

Color Yellow
UN Number 3267
Formula Weight 203.37
Quantity 25 mL
Physical Form Clear Liquid at 20°C
Chemical Name or Material Tetrapropylammonium Hydroxide (20-25% in Water)
EINECSNumber (2)-0186
RTECSNumber BS8400000
TSCA Yes