missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium chloride, 94%

Catalog No. p-3779678
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420710250 25 g Plastic bottle
AC420711000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC420710250 Supplier Thermo Scientific Chemicals Supplier No. 420710250

CAS: 5810-42-4 | C12H28ClN | 221.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 5810-42-4
Molecular Formula C12H28ClN
Molecular Weight (g/mol) 221.81
MDL Number MFCD00038729
InChI Key FBEVECUEMUUFKM-UHFFFAOYSA-M
Synonym tetrapropylammonium chloride, tetrapropyl ammonium chloride, tetra-n-propylammonium chloride, 1-propanaminium, n,n,n-tripropyl-, chloride, 1-propanaminium, n,n,n-tripropyl-, chloride 1:1, n,n,n-tripropylpropan-1-aminium chloride, tetrapropylazanium chloride, acmc-209m2k, ksc491c1h, tetra-n-propylammonium chloride 1g
PubChem CID 79880
IUPAC Name tetrapropylazanium;chloride
SMILES CCC[N+](CCC)(CCC)CCC.[Cl-]

Specifications

Melting Point 240.0°C to 242.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 92% min. (Argentometry)
Packaging Plastic bottle
Linear Formula (CH3CH2CH2)4NCl
Quantity 25 g
Formula Weight 221.81
Percent Purity 94%
Physical Form Crystals
Chemical Name or Material Tetrapropylammonium chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 227-375-5

RUO – Research Use Only