missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylphosphonium Tetra-p-tolylborate 98.0+%, TCI America™

Catalog No. T26035G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26035G 5 g
T260325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. T26035G missing translation for 'mfr' TCI America missing translation for 'supplierNo' T26035G

Chemical Identifiers

CAS 181259-35-8
Molecular Formula C52H48BP
Molecular Weight (g/mol) 714.74
MDL Number MFCD08274957
InChI Key FAYPBNFHFCEUQZ-UHFFFAOYSA-N
PubChem CID 23075127
IUPAC Name tetrakis(4-methylphenyl)boranuide;tetraphenylphosphanium
SMILES [B-](C1=CC=C(C=C1)C)(C2=CC=C(C=C2)C)(C3=CC=C(C=C3)C)C4=CC=C(C=C4)C.C1=CC=C(C=C1)[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 714.74
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraphenylphosphonium Tetra-p-tolylborate