missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylphosphonium iodide, 98+%

Catalog No. AAA1153922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1153922 100 g
AAA1153906 5 g
AAA1153914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1153922 Supplier Thermo Scientific Chemicals Supplier No. A1153922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 2065-67-0 | C24H20IP | 466.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2065-67-0
Molecular Formula C24H20IP
Molecular Weight (g/mol) 466.30
MDL Number MFCD00011917
InChI Key AEFPPQGZJFTXDR-UHFFFAOYSA-M
Synonym tetraphenylphosphonium iodide, tetraphenylphosphoniumiodide, tetraphenylphosphanium iodide, acmc-1cpca
PubChem CID 2724164
IUPAC Name tetraphenylphosphanium;iodide
SMILES [I-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 345°C to 350°C
Quantity 100 g
Beilstein 3922216
Sensitivity Light sensitive
Formula Weight 466.31
Percent Purity ≥98%
Chemical Name or Material Tetraphenylphosphonium iodide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 218-179-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only