missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraphenylphosphonium Iodide 98.0+%, TCI America™
$182.68 - $182.68
Chemical Identifiers
| CAS | 2065-67-0 |
|---|---|
| Molecular Formula | C24H20IP |
| Molecular Weight (g/mol) | 466.30 |
| MDL Number | MFCD00011917 |
| InChI Key | AEFPPQGZJFTXDR-UHFFFAOYSA-M |
| Synonym | tetraphenylphosphonium iodide, tetraphenylphosphoniumiodide, tetraphenylphosphanium iodide, acmc-1cpca |
| PubChem CID | 2724164 |
| IUPAC Name | tetraphenylphosphanium iodide |
| SMILES | [I-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 2065-67-0 | |
| 466.30 | |
| AEFPPQGZJFTXDR-UHFFFAOYSA-M | |
| 2724164 | |
| [I-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C24H20IP | |
| MFCD00011917 | |
| tetraphenylphosphonium iodide, tetraphenylphosphoniumiodide, tetraphenylphosphanium iodide, acmc-1cpca | |
| tetraphenylphosphanium iodide |
Specifications
| 2065-67-0 | |
| C24H20IP | |
| 10 g | |
| AEFPPQGZJFTXDR-UHFFFAOYSA-M | |
| tetraphenylphosphanium iodide | |
| 2724164 | |
| ≥98.0% (HPLC) | |
| Tetraphenylphosphonium Iodide |
| White-Yellow | |
| MFCD00011917 | |
| tetraphenylphosphonium iodide, tetraphenylphosphoniumiodide, tetraphenylphosphanium iodide, acmc-1cpca | |
| [I-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 466.30 | |
| 466.30 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2500
TSCA : No