missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraphenylphosphonium Chloride 98.0+%, TCI America™
$103.97 - $302.25
Chemical Identifiers
| CAS | 2001-45-8 |
|---|---|
| Molecular Formula | C24H20ClP |
| Molecular Weight (g/mol) | 374.85 |
| MDL Number | MFCD00011916 |
| InChI Key | WAGFXJQAIZNSEQ-UHFFFAOYSA-M |
| Synonym | tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co |
| PubChem CID | 164911 |
| IUPAC Name | tetraphenylphosphanium chloride |
| SMILES | [Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 2001-45-8 | |
| 374.85 | |
| WAGFXJQAIZNSEQ-UHFFFAOYSA-M | |
| 164911 | |
| [Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C24H20ClP | |
| MFCD00011916 | |
| tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co | |
| tetraphenylphosphanium chloride |
Specifications
| 2001-45-8 | |
| MFCD00011916 | |
| tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co | |
| [Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 374.85 | |
| 374.85 | |
| Crystalline Powder |
| C24H20ClP | |
| 5 g | |
| WAGFXJQAIZNSEQ-UHFFFAOYSA-M | |
| tetraphenylphosphanium chloride | |
| 164911 | |
| ≥98.0% (HPLC,T) | |
| Tetraphenylphosphonium Chloride |
Safety and Handling
TSCA : No