missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylphosphonium chloride, 98%

Catalog No. AAA1057504
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1057504 2 g
AAA1057509 10 g
AAA1057518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1057504 Supplier Thermo Scientific Chemicals Supplier No. A1057504
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2001-45-8 | C24H20ClP | 374.85 g/mol

Tetraphenylphosphonium chloride is used to generate lipophilic salts from inorganic and organometallic anions. Thus, Ph4P+ is useful as a phase-transfer catalyst, again because it allows inorganic anions to dissolve in organic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraphenylphosphonium chloride is used to generate lipophilic salts from inorganic and organometallic anions. Thus, Ph4P+ is useful as a phase-transfer catalyst, again because it allows inorganic anions to dissolve in organic solvents.

Solubility
Soluble in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2001-45-8
Molecular Formula C24H20ClP
Molecular Weight (g/mol) 374.85
MDL Number MFCD00011916
InChI Key WAGFXJQAIZNSEQ-UHFFFAOYSA-M
Synonym tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co
PubChem CID 164911
IUPAC Name tetraphenylphosphanium;chloride
SMILES [Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 272°C to 278°C
Quantity 2 g
Beilstein 3922393
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 374.86
Percent Purity 98%
Chemical Name or Material Tetraphenylphosphonium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-890-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only