Learn More
Tetraphenylphosphonium bromide, 98+%
CAS: 2751-90-8 | C24H20BrP | 419.30 g/mol
$76.07 - $400.73
Chemical Identifiers
| CAS | 2751-90-8 |
|---|---|
| Molecular Formula | C24H20BrP |
| Molecular Weight (g/mol) | 419.30 |
| MDL Number | MFCD00011915 |
| InChI Key | BRKFQVAOMSWFDU-UHFFFAOYSA-M |
| Synonym | tetraphenylphosphonium bromide, phosphonium, tetraphenyl-, bromide, tetraphenylphosphoniumbromide, tetraphenylphosponium bromide, tetraphenylphosphanium bromide, phosphonium, tetraphenyl-, bromide 1:1, ph4pbr, bromotetraphenylphosphorane, acmc-1ce9k, tetraphenylphosphorus bromide |
| PubChem CID | 2724163 |
| IUPAC Name | tetraphenylphosphanium;bromide |
| SMILES | [Br-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1586009
|
Thermo Scientific Chemicals
A1586009 |
10 g |
Each for $76.07
|
|
|||||
|
AAA1586018
|
Thermo Scientific Chemicals
A1586018 |
50 g |
Each for $222.42
|
|
|||||
|
AAA1586022
|
Thermo Scientific Chemicals
A1586022 |
100 g |
Each for $400.73
|
|
|||||
Description
Tetraphenylphosphonium bromide is a supporting electrolyte for the electroreduction of buckminsterfullerene. It is used to extract heavy metals from aqueous solution as ion-association complexes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetraphenylphosphonium bromide is a supporting electrolyte for the electroreduction of buckminsterfullerene. It is used to extract heavy metals from aqueous solution as ion-association complexes
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Incompatible with strong oxidizing agents. Stable at room temperature in closed containers under normal storage and handling conditions.
Chemical Identifiers
| 2751-90-8 | |
| 419.30 | |
| BRKFQVAOMSWFDU-UHFFFAOYSA-M | |
| 2724163 | |
| [Br-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C24H20BrP | |
| MFCD00011915 | |
| tetraphenylphosphonium bromide, phosphonium, tetraphenyl-, bromide, tetraphenylphosphoniumbromide, tetraphenylphosponium bromide, tetraphenylphosphanium bromide, phosphonium, tetraphenyl-, bromide 1:1, ph4pbr, bromotetraphenylphosphorane, acmc-1ce9k, tetraphenylphosphorus bromide | |
| tetraphenylphosphanium;bromide |
Specifications
| 2751-90-8 | |
| C24H20BrP | |
| 10 g | |
| Hygroscopic | |
| tetraphenylphosphonium bromide, phosphonium, tetraphenyl-, bromide, tetraphenylphosphoniumbromide, tetraphenylphosponium bromide, tetraphenylphosphanium bromide, phosphonium, tetraphenyl-, bromide 1:1, ph4pbr, bromotetraphenylphosphorane, acmc-1ce9k, tetraphenylphosphorus bromide | |
| BRKFQVAOMSWFDU-UHFFFAOYSA-M | |
| tetraphenylphosphanium;bromide | |
| 2724163 | |
| ≥98% |
| 296°C to 302°C | |
| MFCD00011915 | |
| 3922383 | |
| 14,9237 | |
| Soluble in water. | |
| [Br-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 419.30 | |
| 419.31 | |
| Tetraphenylphosphonium bromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 220-393-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only