missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylmethane 96.0+%, TCI America™

Catalog No. T01691G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01691G 1 g
T01695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T01691G Supplier TCI America Supplier No. T01691G

Chemical Identifiers

CAS 630-76-2
Molecular Formula C25H20
Molecular Weight (g/mol) 320.435
MDL Number MFCD00014428
InChI Key PEQHIRFAKIASBK-UHFFFAOYSA-N
Synonym tetraphenylmethane, methane, tetraphenyl, tetraphenyl methane, benzene, 1,1',1,1'-methanetetrayltetrakis, triphenylmethyl benzene, benzene, 1,1',1,1'-methanethetrayltetrakis, acmc-20ajiz, pubchem10677
PubChem CID 12424
IUPAC Name tritylbenzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 283°C
Boiling Point 431°C
Quantity 1 g
Formula Weight 320.44
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetraphenylmethane
RTECSNumber PB4050000
TSCA No
Recommended Storage Refrigerator