Learn More
Tetraphenylmethane, 96%
CAS: 630-76-2 | C25H20 | 320.435 g/mol
Supplier: Thermo Scientific Chemicals A1470606
Description
Tetraphenylmethane is used to prepare a tetrapyridone tecton, which forms a diamondoid network with large internal chambers. It is also used in the preparation of 1-{4-[tris-(4-acetyl-phenyl)-methyl]-phenyl}-ethanone by Friedel-Crafts acylation reaction with acetyl chloride using aluminum chloride as catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetraphenylmethane is used to prepare a tetrapyridone tecton, which forms a diamondoid network with large internal chambers. It is also used in the preparation of 1-{4-[tris-(4-acetyl-phenyl)-methyl]-phenyl}-ethanone by Friedel-Crafts acylation reaction with acetyl chloride using aluminum chloride as catalyst.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
| Tetraphenylmethane | |
| 280°C to 285°C | |
| C25H20 | |
| 5 g | |
| tetraphenylmethane, methane, tetraphenyl, tetraphenyl methane, benzene, 1,1',1,1'-methanetetrayltetrakis, triphenylmethyl benzene, benzene, 1,1',1,1'-methanethetrayltetrakis, acmc-20ajiz, pubchem10677 | |
| PEQHIRFAKIASBK-UHFFFAOYSA-N | |
| tritylbenzene | |
| 12424 | |
| 96% |
| 630-76-2 | |
| 431°C | |
| MFCD00014428 | |
| 1913557 | |
| Insoluble in water. | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4 | |
| 320.435 | |
| 320.44 |
Chemical Identifiers
| 630-76-2 | |
| 320.435 | |
| PEQHIRFAKIASBK-UHFFFAOYSA-N | |
| 12424 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4 |
| C25H20 | |
| MFCD00014428 | |
| tetraphenylmethane, methane, tetraphenyl, tetraphenyl methane, benzene, 1,1',1,1'-methanetetrayltetrakis, triphenylmethyl benzene, benzene, 1,1',1,1'-methanethetrayltetrakis, acmc-20ajiz, pubchem10677 | |
| tritylbenzene |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H351
EINECSNumber : 211-144-0
RTECSNumber : PB4050000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only