missing translation for 'onlineSavingsMsg'
Learn More

Tetrapentylammonium bromide, 99+%

CAS: 866-97-7 | C20H44BrN | 378.48 g/mol

$65.68 - $65.68

Chemical Identifiers

CAS 866-97-7
Molecular Formula C20H44BrN
Molecular Weight (g/mol) 378.48
MDL Number MFCD00011856
InChI Key SPALIFXDWQTXKS-UHFFFAOYSA-M
Synonym tetrapentylammonium bromide, tetraamylammonium bromide, tetra-n-amylammonium bromide, 1-pentanaminium, n,n,n-tripentyl-, bromide 1:1, tetrapentylazanium bromide, n,n,n-tripentylpentan-1-aminium bromide, tetraamylammoniumbromide, 1-pentanaminium, n,n,n-tripentyl-, bromide, acmc-209qb0, tetrapentyl ammonium bromide
PubChem CID 70086
SMILES [Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC221480050
SDS View Documents
Thermo Scientific Chemicals
221480050
5 g Glass bottle
Each for $65.68
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Chemical Identifiers

866-97-7
378.48
SPALIFXDWQTXKS-UHFFFAOYSA-M
70086
C20H44BrN
MFCD00011856
tetrapentylammonium bromide, tetraamylammonium bromide, tetra-n-amylammonium bromide, 1-pentanaminium, n,n,n-tripentyl-, bromide 1:1, tetrapentylazanium bromide, n,n,n-tripentylpentan-1-aminium bromide, tetraamylammoniumbromide, 1-pentanaminium, n,n,n-tripentyl-, bromide, acmc-209qb0, tetrapentyl ammonium bromide
[Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC

Specifications

866-97-7
White
99% min. (Argentometry)
C20H44BrN
MFCD00011856
04, III, 331
SPALIFXDWQTXKS-UHFFFAOYSA-M
tetrapentylazanium bromide
70086
99+%
Tetrapentylammonium bromide
100°C to 102°C
Authentic
Glass bottle
[CH3(CH2)4]4NBr
5 g
tetrapentylammonium bromide, tetraamylammonium bromide, tetra-n-amylammonium bromide, 1-pentanaminium, n,n,n-tripentyl-, bromide 1:1, tetrapentylazanium bromide, n,n,n-tripentylpentan-1-aminium bromide, tetraamylammoniumbromide, 1-pentanaminium, n,n,n-tripentyl-, bromide, acmc-209qb0, tetrapentyl ammonium bromide
[Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC
378.48
378.47
Shiny Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-756-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only