missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetranitro blue tetrazolium chloride, 97%

Catalog No. p-7042024
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61080MC 100 mg
AAJ6108003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61080MC Supplier Thermo Scientific Chemicals Supplier No. J61080MC
Only null left

Redox indicator for enzymes | CAS: 1184-43-6 | C40H28Cl2N12O10 | 907.64 g/mol

Redox indicator for enzymes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1184-43-6
Molecular Formula C40H28Cl2N12O10
Molecular Weight (g/mol) 907.64
MDL Number MFCD00036338
InChI Key VCESGVLABVSDRO-UHFFFAOYSA-L
Synonym TNBT
PubChem CID 129893520
SMILES [Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Beilstein 3897475
IUPAC Name 2-{4'-[3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl]-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl}-3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium dichloride
Formula Weight 999.78
Percent Purity 97%
Color Yellow
Quantity 100 mg
Physical Form Powder
Chemical Name or Material Tetranitro blue tetrazolium chloride
Hazard Category H350
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 214-665-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only