missing translation for 'onlineSavingsMsg'
Learn More

Tetrandrine 98.0+%, TCI America™

Catalog No. p-7137696
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3321200MG 200 mg
T33211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3321200MG Supplier TCI America Supplier No. T3321200MG

Chemical Identifiers

CAS 518-34-3
Molecular Formula C38H42N2O6
Molecular Weight (g/mol) 622.762
MDL Number MFCD08689909
InChI Key WVTKBKWTSCPRNU-KYJUHHDHSA-N
Synonym (1beta)-6,6′C,7,12-Tetramethoxy-2,2′C-dimethylberbaman
PubChem CID 73078
ChEBI CHEBI:49
SMILES CN1CCC2=CC(=C3C=C2C1CC4=CC=C(C=C4)OC5=C(C=CC(=C5)CC6C7=C(O3)C(=C(C=C7CCN6C)OC)OC)OC)OC

Specifications

Melting Point 223°C
Color White-Yellow
Quantity 200 mg
Formula Weight 622.76
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrandrine
RTECSNumber XE9350000
TSCA No