accessibility menu, dialog, popup

UserName

CAS RN 518-34-3

IUPAC Name: (1S,14S)-9,20,21,25-tetramethoxy-15,30-dimethyl-7,23-dioxa-15,30-diazaheptacyclo[22.6.2.2³,⁶.1⁸,¹².1¹⁴,¹⁸.0²⁷,³¹.0²²,³³]hexatriaconta-3,5,8(34),9,11,18,20,22(33),24(32),25,27(31),35-dodecaene
Synonyms: tetrandrine tetrandrine, (1'β)- DL-tetrandrine tetrandrin (S,S)-tetrandrine isotetrandrine tetrandrine, (1'β)-isomer (+)-tetrandrine tetradrine D-tetrandrine
Molecular Weight (g/mol): 622.76
Molecular Formula: C38H42N2O6
InChi Key: WVTKBKWTSCPRNU-KYJUHHDHSA-N
SMILES: COC1=CC=C2C[C@@H]3N(C)CCC4=CC(OC)=C(OC)C(OC5=CC6=C(CCN(C)[C@H]6CC6=CC=C(OC1=C2)C=C6)C=C5OC)=C34
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results