missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TETRAN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116158
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116158 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116158 Supplier Invitrogen™ Supplier No. PA5116158
Only null left

Rabbit Polyclonal Antibody

TETRAN Polyclonal Antibody for Western Blot

TETRAN's overexpression in cultured human cells caused resistance to some NSAIDs (Non-steroidal anti-inflammatory drugs), suggesting that TETRAN is an efflux pump for some NSAIDs.

Specifications

Antigen TETRAN
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MFSD10
Gene Accession No. Q14728
Gene Alias 0610009O03Rik; major facilitator superfamily domain containing 10; major facilitator superfamily domain-containing protein 10; MFSD10; RGD1311900; Tetracycline transporter-like protein; TETRAN; TETTRAN
Gene Symbols MFSD10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 245-274 amino acids from the Central region of human MFS10.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10227
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less