missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylthiourea 98.0+%, TCI America™

Catalog No. T015625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T015625G 25 g
T0156100G 100 g
T0156500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T015625G Supplier TCI America Supplier No. T015625G

Chemical Identifiers

CAS 2782-91-4
Molecular Formula C5H12N2S
Molecular Weight (g/mol) 132.23
MDL Number MFCD00008324
InChI Key MNOILHPDHOHILI-UHFFFAOYSA-N
Synonym tetramethylthiourea, thiourea, tetramethyl, basthioryl, 1,1,3,3-tetramethyl-2-thiourea, tmtu, n,n,n',n'-tetramethylthiourea, urea, thio-, tetramethyl, tetramethyl-2-thiourea, urea, 1,1,3,3-tetramethyl-2-thio, thiourea, n,n,n',n'-tetramethyl
PubChem CID 17725
IUPAC Name 1,1,3,3-tetramethylthiourea
SMILES CN(C)C(=S)N(C)C

Specifications

Melting Point 80°C
Color White
Boiling Point 243°C
Quantity 25 g
Formula Weight 132.23
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylthiourea
EINECSNumber (2)-2868
RTECSNumber YU2750000
TSCA Yes