missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylsilane 99.0+%, TCI America™
Supplier: TCI America T0154100ML
Specifications
| Tetramethylsilane [for NMR] | |
| Colorless | |
| C4H12Si | |
| 100 mL | |
| silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073 | |
| C[Si](C)(C)C | |
| 88.23 | |
| CHEBI:85361 | |
| ≥99.0% (GC) |
| 75-76-3 | |
| 26°C | |
| MFCD00008274 | |
| 2749 | |
| CZDYPVPMEAXLPK-UHFFFAOYSA-N | |
| tetramethylsilane | |
| 6396 | |
| 88.23 | |
| Liquid |
Chemical Identifiers
| 75-76-3 | |
| 88.23 | |
| CZDYPVPMEAXLPK-UHFFFAOYSA-N | |
| 6396 | |
| tetramethylsilane |
| C4H12Si | |
| MFCD00008274 | |
| silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073 | |
| CHEBI:85361 | |
| C[Si](C)(C)C |
Safety and Handling
EINECSNumber : (2)-0660
TSCA : Yes
Recommended Storage : Refrigerator