missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylsilane 99.0+%, TCI America™

Catalog No. p-7136111
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T015425ML 25 mL
T0154100ML 100 mL
T0154500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T015425ML Supplier TCI America Supplier No. T015425ML
Only null left

Chemical Identifiers

CAS 75-76-3
Molecular Formula C4H12Si
Molecular Weight (g/mol) 88.23
MDL Number MFCD00008274
InChI Key CZDYPVPMEAXLPK-UHFFFAOYSA-N
Synonym silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073
PubChem CID 6396
ChEBI CHEBI:85361
IUPAC Name tetramethylsilane
SMILES C[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 26°C
Quantity 25 mL
UN Number 2749
Formula Weight 88.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetramethylsilane [for NMR]
EINECSNumber (2)-0660
TSCA Yes
Recommended Storage Refrigerator