missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tetramethylrhodamine Cadaverine, 5-(and-6)-((N-(5-Aminopentyl) Amino) Carbonyl) Tetramethylrhodamine, mixed isomers

Catalog No. A1318
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1318 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1318 Supplier Invitrogen™ Supplier No. LSA1318

Tetramethylrhodamine Cadaverine, 5-(and-6)-((N-(5-Aminopentyl) Amino) Carbonyl) Tetramethylrhodamine, mixed isomers

The primary aliphatic amine of tetramethylrhodamine cadaverine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247). This material consists of the 5- and 6- isomer. Please see the structure for additional details.

Specifications

Quantity 10 mg
Product Type Tetramethylrhodamine Cadaverine
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Shipping Condition Room Temperature
Reactive Moiety Amine, Cadaverine
Label or Dye TMR (Tetramethylrhodamine)
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.