missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tetramethylrhodamine-5-(and-6)-Isothiocyanate (5(6)-TRITC), mixed isomers

Catalog No. t490
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T490 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T490 Supplier Invitrogen™ Supplier No. T490

Tetramethylrhodamine-5-(and-6)-Isothiocyanate (5(6)-TRITC), mixed isomers

The thiol-reactive tetramethylrhodamine-5-(and-6)-isothiocyanate (5(6)-TRITC) can be used to can be used to create bright orange-red-fluorescent bioconjugates with excitation/emission maxima ∼555/580.

Specifications

Quantity 10 mg
Product Type Tetramethylrhodamine-5-(and-6)-Isothiocyanate
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Isothiocyanate
Label or Dye TMR (Tetramethylrhodamine)
Label Type Classic Dyes
Emission 580
Excitation 555
What is the excitation and emission wavelength for rhodamine?

Rhodamine is a generic term for a wide variety of cationic dyes whose fluorescence emission can range from green, orange to red. The table below lists the excitation and emission maxima (nm), as well as molar extinction coefficients (“EC”; cm-1 M-1), for various rhodamine dyes (data derived with dye dissolved in methanol).

Dye Excitation Emission EC
Rhodamine B 568 583 88,000
Rhodamine 123 507 529 101,000
Rhodamine 110 499 521 92,000
Rhodamine 6G 528 551 105,000
XRITC 572 596 92,000

For Research Use Only. Not for use in diagnostic procedures.