missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylphosphonium bromide, 97%

Catalog No. AAA1790703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1790703 1 g
AAA1790706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1790703 Supplier Thermo Scientific Chemicals Supplier No. A1790703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4519-28-2 | C4H12BrP | 171.02 g/mol

It is used as a pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate and in chemical research.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 4519-28-2
Molecular Formula C4H12BrP
Molecular Weight (g/mol) 171.02
MDL Number MFCD00011802
InChI Key ZTXFOCMYRCGSMU-UHFFFAOYSA-M
Synonym tetramethylphosphonium bromide, tetramethylphosphanium bromide, acmc-1ai8x, ch3 4pbr, tetramethyl phosphonium bromide, phosphonium, tetramethyl-, bromide
PubChem CID 357594
IUPAC Name tetramethylphosphanium;bromide
SMILES [Br-].C[P+](C)(C)C

Specifications

Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 171.03
Percent Purity 98%
Chemical Name or Material Tetramethylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only