accessibility menu, dialog, popup

UserName

CAS RN 4519-28-2

IUPAC Name: tetramethylphosphanium bromide
Synonyms: tetramethylphosphanium bromide
Molecular Weight (g/mol): 171.02
Molecular Formula: C4H12BrP
InChi Key: ZTXFOCMYRCGSMU-UHFFFAOYSA-M
SMILES: [Br-].C[P+](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results