missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylene Sulfoxide 98.0+%, TCI America™

Catalog No. p-7136107
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T014625G 25 g
T0146500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T014625G Supplier TCI America Supplier No. T014625G
Only null left

Chemical Identifiers

CAS 1600-44-8
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00005477
InChI Key ISXOBTBCNRIIQO-UHFFFAOYSA-N
Synonym tetramethylene sulfoxide, tetrahydrothiophene 1-oxide, thiophane oxide, tetrahydrothiophene oxide, thiophane monoxide, thiophane 1-oxide, thiophene, tetrahydro-, 1-oxide, tetramethylene sulphoxide, thiophene, tetrahydro, 1-oxide, tetrametylene sulfoxide
PubChem CID 1128
IUPAC Name thiolane 1-oxide
SMILES C1CCS(=O)C1

Specifications

Color Yellow
Boiling Point 107°C
Quantity 25 g
Formula Weight 104.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetramethylene Sulfoxide
RTECSNumber XN0830000
TSCA Yes