missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylcyclobutane-1,3-dione, 99%

CAS: 933-52-8 | C8H12O2 | 140.182 g/mol

Supplier:  Thermo Scientific Chemicals A1629718

 View more versions of this product

Catalog No. AAA1629718


Only null left
Explore available promotions

Description

Description

Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.

Solubility
It is soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
Specifications

Specifications

Tetramethylcyclobutane-1,3-dione
112°C to 115°C
C8H12O2
50 g
It is soluble in chloroform.
CC1(C(=O)C(C1=O)(C)C)C
140.182
140.18
933-52-8
120°C to 122°C (150 mmHg)
MFCD00001331
tetramethyl-1,3-cyclobutanedione, tetramethylcyclobutane-1,3-dione, 1,3-cyclobutanedione, 2,2,4,4-tetramethyl, tetramethylcyclobuta-1,3-dione, 2,2,4,4-tetramethyl-1,3-cyclobutanedione, unii-rt4aq22ks4, 1,1,3,3-tetramethylcyclobutanedione, 2,2,4,4-tetramethylcyclobutanedione, dimethyl ketene dimer, rt4aq22ks4
RGCDVHNITQEYPO-UHFFFAOYSA-N
2,2,4,4-tetramethylcyclobutane-1,3-dione
13617
99%

Chemical Identifiers

933-52-8
140.182
RGCDVHNITQEYPO-UHFFFAOYSA-N
13617
CC1(C(=O)C(C1=O)(C)C)C
C8H12O2
MFCD00001331
tetramethyl-1,3-cyclobutanedione, tetramethylcyclobutane-1,3-dione, 1,3-cyclobutanedione, 2,2,4,4-tetramethyl, tetramethylcyclobuta-1,3-dione, 2,2,4,4-tetramethyl-1,3-cyclobutanedione, unii-rt4aq22ks4, 1,1,3,3-tetramethylcyclobutanedione, 2,2,4,4-tetramethylcyclobutanedione, dimethyl ketene dimer, rt4aq22ks4
2,2,4,4-tetramethylcyclobutane-1,3-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 213-269-6

RTECSNumber : GU1775000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only