accessibility menu, dialog, popup

UserName

CH3H3COCH3H3CO

CAS RN 933-52-8

IUPAC Name: tetramethylcyclobutane-1,3-dione
Synonyms: tetramethylcyclobutane-1,3-dione
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: RGCDVHNITQEYPO-UHFFFAOYSA-N
SMILES: CC1(C)C(=O)C(C)(C)C1=O
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)