missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylcyclobutane-1,3-dione, 99%

Catalog No. AAA1629709
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1629709 10 g
AAA1629718 50 g
AAA1629730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1629709 Supplier Thermo Scientific Chemicals Supplier No. A1629709

CAS: 933-52-8 | C8H12O2 | 140.182 g/mol

Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.

Solubility
It is soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.

Chemical Identifiers

CAS 933-52-8
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00001331
InChI Key RGCDVHNITQEYPO-UHFFFAOYSA-N
Synonym tetramethyl-1,3-cyclobutanedione, tetramethylcyclobutane-1,3-dione, 1,3-cyclobutanedione, 2,2,4,4-tetramethyl, tetramethylcyclobuta-1,3-dione, 2,2,4,4-tetramethyl-1,3-cyclobutanedione, unii-rt4aq22ks4, 1,1,3,3-tetramethylcyclobutanedione, 2,2,4,4-tetramethylcyclobutanedione, dimethyl ketene dimer, rt4aq22ks4
PubChem CID 13617
IUPAC Name 2,2,4,4-tetramethylcyclobutane-1,3-dione
SMILES CC1(C(=O)C(C1=O)(C)C)C

Specifications

Melting Point 112°C to 115°C
Boiling Point 120°C to 122°C (150 mmHg)
Quantity 10 g
Solubility Information It is soluble in chloroform.
Formula Weight 140.18
Percent Purity 99%
Chemical Name or Material Tetramethylcyclobutane-1,3-dione
EINECSNumber 213-269-6
RTECSNumber GU1775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only