missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium tetrafluoroborate, 97%

CAS: 661-36-9 | C4H12BF4N | 160.95 g/mol

$58.62 - $420.14

Chemical Identifiers

CAS 661-36-9
Molecular Formula C4H12BF4N
Molecular Weight (g/mol) 160.95
MDL Number MFCD00011745
InChI Key XWFABLFRYCHILB-UHFFFAOYSA-N
Synonym tetramethylammonium tetrafluoroborate, tetramethylammonium fluoroborate, borate 1-, tetrafluoro-, tetramethylammonium salt, tetramethylazanium tetrafluoroborate, n,n,n-trimethylmethanaminium tetrafluoroborate 1-, tetramethylammonium ion tetrafluoroborate, ammonium, tetramethyl-, tetrafluoroborate 1-, acmc-209ntt, methanaminium, n,n,n-trimethyl-, tetrafluoroborate 1-, tetramethyl ammonium tetrafluoroborate
PubChem CID 12621
IUPAC Name tetramethylazanium;tetrafluoroborate
SMILES C[N+](C)(C)C.F[B-](F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1743606
View Documents
Thermo Scientific Chemicals
A1743606
5 g
Each for $58.62
Only null left
 
AAA1743614
View Documents
Thermo Scientific Chemicals
A1743614
25 g
Each for $148.45
Only null left
 
AAA1743622
View Documents
Thermo Scientific Chemicals
A1743622
100 g
Each for $420.14
Only null left
 
Description

Description

It is used in the catalytic reduction of 1,6-dihalohexanes by nickel(i) salen electrogenerated at glassy carbon cathodes in dimethylformamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the catalytic reduction of 1,6-dihalohexanes by nickel(i) salen electrogenerated at glassy carbon cathodes in dimethylformamide.

Solubility
Solubility in hot water is almost transparent.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
Specifications

Chemical Identifiers

661-36-9
160.95
XWFABLFRYCHILB-UHFFFAOYSA-N
12621
C[N+](C)(C)C.F[B-](F)(F)F
C4H12BF4N
MFCD00011745
tetramethylammonium tetrafluoroborate, tetramethylammonium fluoroborate, borate 1-, tetrafluoro-, tetramethylammonium salt, tetramethylazanium tetrafluoroborate, n,n,n-trimethylmethanaminium tetrafluoroborate 1-, tetramethylammonium ion tetrafluoroborate, ammonium, tetramethyl-, tetrafluoroborate 1-, acmc-209ntt, methanaminium, n,n,n-trimethyl-, tetrafluoroborate 1-, tetramethyl ammonium tetrafluoroborate
tetramethylazanium;tetrafluoroborate

Specifications

661-36-9
C4H12BF4N
5 g
Hygroscopic
XWFABLFRYCHILB-UHFFFAOYSA-N
tetramethylazanium;tetrafluoroborate
12621
97%
417°C to 419°C
MFCD00011745
3715404
tetramethylammonium tetrafluoroborate, tetramethylammonium fluoroborate, borate 1-, tetrafluoro-, tetramethylammonium salt, tetramethylazanium tetrafluoroborate, n,n,n-trimethylmethanaminium tetrafluoroborate 1-, tetramethylammonium ion tetrafluoroborate, ammonium, tetramethyl-, tetrafluoroborate 1-, acmc-209ntt, methanaminium, n,n,n-trimethyl-, tetrafluoroborate 1-, tetramethyl ammonium tetrafluoroborate
C[N+](C)(C)C.F[B-](F)(F)F
160.95
160.95
Tetramethylammonium tetrafluoroborate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-547-1

RTECSNumber : BS8300000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only