Learn More
Tetramethylammonium tetrafluoroborate, 97%
CAS: 661-36-9 | C4H12BF4N | 160.95 g/mol
Supplier: Thermo Scientific Chemicals A1743622
Description
It is used in the catalytic reduction of 1,6-dihalohexanes by nickel(i) salen electrogenerated at glassy carbon cathodes in dimethylformamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the catalytic reduction of 1,6-dihalohexanes by nickel(i) salen electrogenerated at glassy carbon cathodes in dimethylformamide.
Solubility
Solubility in hot water is almost transparent.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
Specifications
| Tetramethylammonium tetrafluoroborate | |
| 417°C to 419°C | |
| MFCD00011745 | |
| 3715404 | |
| tetramethylammonium tetrafluoroborate, tetramethylammonium fluoroborate, borate 1-, tetrafluoro-, tetramethylammonium salt, tetramethylazanium tetrafluoroborate, n,n,n-trimethylmethanaminium tetrafluoroborate 1-, tetramethylammonium ion tetrafluoroborate, ammonium, tetramethyl-, tetrafluoroborate 1-, acmc-209ntt, methanaminium, n,n,n-trimethyl-, tetrafluoroborate 1-, tetramethyl ammonium tetrafluoroborate | |
| C[N+](C)(C)C.F[B-](F)(F)F | |
| 160.95 | |
| 160.95 |
| 661-36-9 | |
| C4H12BF4N | |
| 100 g | |
| Hygroscopic | |
| XWFABLFRYCHILB-UHFFFAOYSA-N | |
| tetramethylazanium;tetrafluoroborate | |
| 12621 | |
| 97% |
Chemical Identifiers
| 661-36-9 | |
| 160.95 | |
| XWFABLFRYCHILB-UHFFFAOYSA-N | |
| 12621 | |
| C[N+](C)(C)C.F[B-](F)(F)F |
| C4H12BF4N | |
| MFCD00011745 | |
| tetramethylammonium tetrafluoroborate, tetramethylammonium fluoroborate, borate 1-, tetrafluoro-, tetramethylammonium salt, tetramethylazanium tetrafluoroborate, n,n,n-trimethylmethanaminium tetrafluoroborate 1-, tetramethylammonium ion tetrafluoroborate, ammonium, tetramethyl-, tetrafluoroborate 1-, acmc-209ntt, methanaminium, n,n,n-trimethyl-, tetrafluoroborate 1-, tetramethyl ammonium tetrafluoroborate | |
| tetramethylazanium;tetrafluoroborate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-547-1
RTECSNumber : BS8300000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only