missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium Fluoride Tetrahydrate 98.0+%, TCI America™
Supplier: TCI America T275425G
Specifications
| Tetramethylammonium Fluoride Tetrahydrate | |
| 38°C | |
| C4H20FNO4 | |
| 25 g | |
| HQFTZNVQVRRDLN-UHFFFAOYSA-M | |
| tetramethylazanium;fluoride;tetrahydrate | |
| 2723808 | |
| ≥98.0% (T) |
| 17787-40-5 | |
| White | |
| MFCD00149968 | |
| tetramethylammonium fluoride tetrahydrate, tetramethylazanium fluoride tetrahydrate, methanaminium, n,n,n-trimethyl-, fluoride, tetrahydrate, tetramethylammonium tetrahydrate fluoride, tetramethylammonium ion tetrahydrate fluoride, acmc-1c8r1, tetramethylammoniumfluoridetetrahydrate, methanaminium,n,n,n-trimethyl-,fluoride,tetrahydrate 9ci, tetramethylamine, fluoride, hydrate, hydrate, hydrate, hydrate, methanaminium,n,n,n-trimethyl-, fluoride, tetrahydrate 9ci | |
| C[N+](C)(C)C.O.O.O.O.[F-] | |
| 165.205 | |
| 93.15 | |
| Crystalline Lumps |
Chemical Identifiers
| 17787-40-5 | |
| 165.205 | |
| HQFTZNVQVRRDLN-UHFFFAOYSA-M | |
| 2723808 | |
| C[N+](C)(C)C.O.O.O.O.[F-] |
| C4H20FNO4 | |
| MFCD00149968 | |
| tetramethylammonium fluoride tetrahydrate, tetramethylazanium fluoride tetrahydrate, methanaminium, n,n,n-trimethyl-, fluoride, tetrahydrate, tetramethylammonium tetrahydrate fluoride, tetramethylammonium ion tetrahydrate fluoride, acmc-1c8r1, tetramethylammoniumfluoridetetrahydrate, methanaminium,n,n,n-trimethyl-,fluoride,tetrahydrate 9ci, tetramethylamine, fluoride, hydrate, hydrate, hydrate, hydrate, methanaminium,n,n,n-trimethyl-, fluoride, tetrahydrate 9ci | |
| tetramethylazanium;fluoride;tetrahydrate |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator