accessibility menu, dialog, popup

UserName

CH3NCH3CH3H3CH2OH2OH2OH2OF

CAS RN 17787-40-5

IUPAC Name: tetramethylazanium tetrahydrate fluoride
Synonyms: tetramethylammonium ion tetrahydrate fluoride
Molecular Weight (g/mol): 165.21
Molecular Formula: C4H20FNO4
InChi Key: HQFTZNVQVRRDLN-UHFFFAOYSA-M
SMILES: O.O.O.O.[F-].C[N+](C)(C)C
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (5)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)