missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium Chloride 98.0+%, TCI America™
$36.57 - $109.74
Chemical Identifiers
| CAS | 75-57-0 |
|---|---|
| Molecular Formula | C4H12ClN |
| Molecular Weight (g/mol) | 109.597 |
| MDL Number | MFCD00011628 |
| InChI Key | OKIZCWYLBDKLSU-UHFFFAOYSA-M |
| Synonym | tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl |
| PubChem CID | 6379 |
| ChEBI | CHEBI:7070 |
| IUPAC Name | tetramethylazanium;chloride |
| SMILES | C[N+](C)(C)C.[Cl-] |
Chemical Identifiers
| 75-57-0 | |
| 109.597 | |
| OKIZCWYLBDKLSU-UHFFFAOYSA-M | |
| 6379 | |
| tetramethylazanium;chloride |
| C4H12ClN | |
| MFCD00011628 | |
| tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl | |
| CHEBI:7070 | |
| C[N+](C)(C)C.[Cl-] |
Specifications
| 75-57-0 | |
| C4H12ClN | |
| 25 g | |
| tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl | |
| C[N+](C)(C)C.[Cl-] | |
| 109.597 | |
| CHEBI:7070 | |
| ≥98.0% (T) | |
| Tetramethylammonium Chloride |
| White | |
| MFCD00011628 | |
| 2811 | |
| OKIZCWYLBDKLSU-UHFFFAOYSA-M | |
| tetramethylazanium;chloride | |
| 6379 | |
| 109.60 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0186
RTECSNumber : BS7700000
TSCA : Yes