missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Chloride 98.0+%, TCI America™

Catalog No. T013625G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T013625G 25 g
T0136500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. T013625G missing translation for 'mfr' TCI America missing translation for 'supplierNo' T013625G

Chemical Identifiers

CAS 75-57-0
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.597
MDL Number MFCD00011628
InChI Key OKIZCWYLBDKLSU-UHFFFAOYSA-M
Synonym tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl
PubChem CID 6379
ChEBI CHEBI:7070
IUPAC Name tetramethylazanium;chloride
SMILES C[N+](C)(C)C.[Cl-]

Specifications

Color White
Quantity 25 g
UN Number 2811
Formula Weight 109.60
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium Chloride
EINECSNumber (2)-0186
RTECSNumber BS7700000
TSCA Yes