missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium chloride, 97%

CAS: 75-57-0 | C4H12ClN | 109.597 g/mol

Supplier:  Thermo Scientific Chemicals A132880B

 View more versions of this product

Catalog No. AAA132880B

Only null left
Explore available promotions

Description

Description

Tetramethylammonium binds AT-rich DNA polymers while concomitantly abolishing the preferential melting of AT versus GC base pairs. Supplied as 0.2 ?m filtered solution in 18 megohm water. It is employed as a phase transfer catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylammonium binds AT-rich DNA polymers while concomitantly abolishing the preferential melting of AT versus GC base pairs. Supplied as 0.2 μm filtered solution in 18 megohm water. It is employed as a phase transfer catalyst.

Solubility
Soluble in water. Insoluble in ether, and benzene.

Notes
Hygroscopic. Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
Specifications

Specifications

Tetramethylammonium chloride
>300°C
Odorless
C4H12ClN
MFCD00011628
UN2811
Hygroscopic
Soluble in water. Insoluble in ether,and benzene.
C[N+](C)(C)C.[Cl-]
109.597
CHEBI:7070
97%
75-57-0
1.17
may cont. ≈3% silica as anticaking agent
(CH3)4NCl
1000 g
2496575
tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl
OKIZCWYLBDKLSU-UHFFFAOYSA-M
tetramethylazanium;chloride
6379
109.6

Chemical Identifiers

75-57-0
109.597
OKIZCWYLBDKLSU-UHFFFAOYSA-M
6379
tetramethylazanium;chloride
C4H12ClN
MFCD00011628
tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl
CHEBI:7070
C[N+](C)(C)C.[Cl-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P307+P311-P312-P330-P332+P313-P361-P363-P501c

H300-H311-H315-H319-H370

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 200-880-8

RTECSNumber : BS7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only