Learn More
Tetramethylammonium acetate, tech. 90%
Description
Tetramethylammonium acetate is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. Further, it is used to prepare (1R,4S)-4-hydroxycyclopent-2-en-1-essigsaeure-methylester by reacting with 1,3-dimethyl-imidazolidin-2-one. In addition to this, it serves as an emulsifier, catalyst, sterilizer, antistatic and antiseptic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tetramethylammonium acetate is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. Further, it is used to prepare (1R,4S)-4-hydroxycyclopent-2-en-1-essigsaeure-methylester by reacting with 1,3-dimethyl-imidazolidin-2-one. In addition to this, it serves as an emulsifier, catalyst, sterilizer, antistatic and antiseptic agent.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
Specifications
Specifications
| Melting Point | 256°C (decomposition) |
| Linear Formula | (CH3)4N(OCOCH3) |
| Quantity | 5 g |
| Beilstein | 3571830 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water. |
| Formula Weight | 133.2 |
| Percent Purity | 90% |
| Grade | Technical |
| Chemical Name or Material | Tetramethylammonium acetate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 234-179-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only