accessibility menu, dialog, popup

UserName

CAS RN 10581-12-1

IUPAC Name: tetramethylazanium acetate
Synonyms: tetramethylammonium ion acetate
Molecular Weight (g/mol): 133.19
Molecular Formula: C6H15NO2
InChi Key: MRYQZMHVZZSQRT-UHFFFAOYSA-M
SMILES: CC([O-])=O.C[N+](C)(C)C
Quantity 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)