missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium acetate, tech. 90%

Catalog No. AAH2765914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2765914 25 g
AAH2765906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2765914 Supplier Thermo Scientific Chemicals Supplier No. H2765914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10581-12-1 | C6H15NO2 | 133.19 g/mol

Tetramethylammonium acetate is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. Further, it is used to prepare (1R,4S)-4-hydroxycyclopent-2-en-1-essigsaeure-methylester by reacting with 1,3-dimethyl-imidazolidin-2-one. In addition to this, it serves as an emulsifier, catalyst, sterilizer, antistatic and antiseptic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylammonium acetate is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. Further, it is used to prepare (1R,4S)-4-hydroxycyclopent-2-en-1-essigsaeure-methylester by reacting with 1,3-dimethyl-imidazolidin-2-one. In addition to this, it serves as an emulsifier, catalyst, sterilizer, antistatic and antiseptic agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.

Chemical Identifiers

CAS 10581-12-1
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00011630
InChI Key MRYQZMHVZZSQRT-UHFFFAOYSA-M
Synonym tetramethylammonium acetate, methanaminium, n,n,n-trimethyl-, acetate, tetramethyl ammonium acetate, methanaminium, n,n,n-trimethyl-, acetate 1:1, tetramethylammonium ion acetate, ammonium, tetramethyl-, acetate, tetramethylazanium acetate, n,n,n-trimethyl-methanaminiuacetate, acetate ion; tetramethylammonium ion
PubChem CID 82741
IUPAC Name tetramethylazanium acetate
SMILES CC([O-])=O.C[N+](C)(C)C

Specifications

Melting Point 256°C (decomposition)
Linear Formula (CH3)4N(OCOCH3)
Quantity 25 g
Beilstein 3571830
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 133.2
Percent Purity 90%
Grade Technical
Chemical Name or Material Tetramethylammonium acetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-179-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only