missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Acetate 98.0+%, TCI America™

Catalog No. p-7136572
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10485G 5 g
T104825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10485G Supplier TCI America Supplier No. T10485G
Only null left

Chemical Identifiers

CAS 10581-12-1
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00011630
InChI Key MRYQZMHVZZSQRT-UHFFFAOYSA-M
Synonym tetramethylammonium acetate, methanaminium, n,n,n-trimethyl-, acetate, tetramethyl ammonium acetate, methanaminium, n,n,n-trimethyl-, acetate 1:1, tetramethylammonium ion acetate, ammonium, tetramethyl-, acetate, tetramethylazanium acetate, n,n,n-trimethyl-methanaminiuacetate, acetate ion; tetramethylammonium ion
PubChem CID 82741
IUPAC Name tetramethylazanium acetate
SMILES CC([O-])=O.C[N+](C)(C)C

Specifications

Melting Point 195°C
Color White
Quantity 5 g
Formula Weight 133.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium Acetate
EINECSNumber (2)-0186
TSCA Yes
Recommended Storage Refrigerator