missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyl-1,3-cyclobutanedione 99.0+%, TCI America™

Catalog No. D02585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D02585G 5 g
D025825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D02585G Supplier TCI America Supplier No. D02585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 933-52-8
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00001331
InChI Key RGCDVHNITQEYPO-UHFFFAOYSA-N
Synonym tetramethyl-1,3-cyclobutanedione, tetramethylcyclobutane-1,3-dione, 1,3-cyclobutanedione, 2,2,4,4-tetramethyl, tetramethylcyclobuta-1,3-dione, 2,2,4,4-tetramethyl-1,3-cyclobutanedione, unii-rt4aq22ks4, 1,1,3,3-tetramethylcyclobutanedione, 2,2,4,4-tetramethylcyclobutanedione, dimethyl ketene dimer, rt4aq22ks4
PubChem CID 13617
IUPAC Name 2,2,4,4-tetramethylcyclobutane-1,3-dione
SMILES CC1(C(=O)C(C1=O)(C)C)C

Specifications

Melting Point 114°C
Color White-Yellow
Boiling Point 159°C
Quantity 5 g
Formula Weight 140.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethyl-1,3-cyclobutanedione [Precursor to Dimethyl Ketene]
RTECSNumber GU1775000
TSCA Yes