missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Tetralone, 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 94.0 |
| CAS Max % | 100.0 |
| Melting Point | 18.0°C |
| Color | Yellow |
| Density | 1.1060g/mL |
| Boiling Point | 131.0°C to 133.0°C (11.0 mmHg) |
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (GC) |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minu |
| EINECSNumber | 208-498-3 |
| TSCA | TSCA |
RUO – Research Use Only