missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(triphenylphosphine)platinum(0), 98%

Catalog No. AC223960010
missing translation for 'changeView'
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223960010 1 g Glass bottle
AC223960050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AC223960010 Supplier Thermo Scientific Chemicals missing translation for 'supplierNo' 223960010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14221-02-4 | C72H60P4Pt | 1244.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14221-02-4
Molecular Formula C72H60P4Pt
Molecular Weight (g/mol) 1244.25
MDL Number MFCD00010014
InChI Key SYKXNRFLNZUGAJ-UHFFFAOYSA-N
Synonym tetrakis triphenylphosphine platinum 0, tetrakis triphenylphosphine platinum, platinum; triphenylphosphane, pt pph3 4, platinum tetratriphenylphosphine, platinum, tetrakis triphenylphosphine-, t-4, platinum 0-tetrakis triphenylphosphine, platinum; triphenylphosphine, tetrakis triphenylphosphine platinum 0 1g
PubChem CID 11979705
IUPAC Name platinum;triphenylphosphane
SMILES [Pt].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 159.0°C to 160.0°C
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula [(C6H5)3P]4Pt
Quantity 1 g
Fieser 15,300; 16,98; 04,33; 05,34; 09,45; 10,33; 14,36; 15,30; 16,29; 17,32; 06,570; 07,357; 09,450; 12,467; 13,294; 16,317
Solubility Information Solubility in water: insoluble.
Formula Weight 1244.27
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetrakis(triphenylphosphine)platinum(0)
Show More Show Less
EINECSNumber 238-087-4

RUO – Research Use Only